Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTPN5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DU8
75931-386EA 518 USD InStock
75931-386
Anti-PTPN5 Rabbit Polyclonal Antibody
Antibodies
The protein tyrosine phosphatase PTPN5, also known as Striatal enriched phosphatase (STEP), is involved in the regulation of synaptic plasticity and neuronal cell survival, including MAPKs, Src family kinases and NMDA receptors (1). It is expressed in dopaminoceptive neurons of the central nervous system and multiple forms of PTPN5 show differential enrichment in adult brain regions (2). NMDA-mediated activation of PTPN5 is an important mechanism for regulation of Erk activity in neurons (3). Furthermore, PTPN5 is involved in the regulation of both NMDAR and AMPAR trafficking (4,5). PTPN5 may play a role in Alzheimer's disease (1).

Anti-PTPN5 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 67 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PTPN5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty