Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-QKI-5 Mouse Monoclonal Antibody (FITC) [Clone: S147-6]

Supplier: Rockland Immunochemicals
200-302-W79
75931-416EA 539 USD InStock
75931-416
Anti-QKI-5 Mouse Monoclonal Antibody (FITC) [Clone: S147-6]
Antibodies
QKI is also called Protein Quaking or HqkI. QKI is an RNA-binding protein that plays a central role in myelinization. QKI acts by regulating pre-mRNA splicing, mRNA export, mRNA stability and protein translation, and is itself, regulated by alternative splicing. QKI is expressed in the frontal cortex of brain, but is shown to be downregulated in the brain of schizophrenic patients. Anti-QKI (pan) is ideal for research in Neuroscience and Gene Expression.

Anti-QKI (pan) FITC Conjugated Antibody is suitable for Western Blots, Immunohistochemistry and Immunocytochemistry. Expect a band approximately ~36-38kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: QKI-5
Clonality: Monoclonal
Clone: S147-6
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty