Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CRADD Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DV9
75931-432EA 518 USD InStock
75931-432
Anti-CRADD Rabbit Polyclonal Antibody
Antibodies
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF and Fas ligand of the TNF family through their death domain (DD)-containing receptors, TNFR1 and Fas. The death signals are transduced by a group of DD-containing adapter molecules. A novel cell death adapter was recently identified by two independent groups and designated RAIDD (RIP-associated ICH-1/CED-3-homologous protein with DD) and CRADD (caspase and RIP adapter with DD)1, RAIDD contains a DD and a CARD (for caspase recruitment domain) which interact with RIP and caspase, respectively, to transduce death signals1, 3. RAIDD is constitutively expressed in many tissues and mediates apoptosis caused by Fas and TNFR-1.

Anti-RAIDD Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 23 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: CRADD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty