Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Rptor Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DW4
75931-456EA 518 USD InStock
75931-456
Anti-Rptor Rabbit Polyclonal Antibody
Antibodies
The mammalian Target of Rapamycin (TOR, also known as mTOR) is an evolutionarily conserved serine/threonine kinase that regulates cell growth and cell cycle through its ability to integrate signals from nutrient levels and growth factors. Rapamycin inhibits TOR activity resulting in reduced cell growth and reduced rates of cell cycle and cell proliferation. Raptor (regulatory associated protein of TOR) is a TOR-binding protein essential for TOR signaling in vivo. It acts as a TOR scaffold protein whose binding by TOR substrates is necessary for effective TOR-catalyzed phosphorylation. These substrates include the ribosomal protein S6 kinase (RP S6K) and the eukaryotic initiation factor 4E binding protein 4EBP1, proteins necessary for cell growth and proliferation and responsive to nutrient and mitogen levels. Raptor binds these proteins through a common 5 amino acid TOR-signaling (TOS) motif; mutation of this motif prevents the TOR-dependent phosphorylation of these proteins.

Anti-Raptor Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 149 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: Rptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty