Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RBBP8 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DW9
75931-484EA 518 USD InStock
75931-484
Anti-RBBP8 Rabbit Polyclonal Antibody
Antibodies
RBBP8, also known as CtBP (carboxy-terminal binding protein) interacting protein (CTIP), was characterized for its role in transcription as a cofactor for the transcriptional repressor CtBP, and also as a binding partner for other proteins including the cell cycle regulators retinoblastoma protein (Rb) and breast cancer 1 (BRCA1). It is ubiquitously expressed and localizes to the nucleus. RBBP8 is thought to modulate the functions in cell proliferation, transcriptional regulation and DNA repair. RBBP8 also plays a central role in the cell cycle checkpoint response to DNA double-stranded breaks (DSBs), with new evidence demonstrating that it controls the choice of DSB repair pathway.

Anti-RBBP8 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 102 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: RBBP8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty