Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RBPMS Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X23
75931-490EA 518 USD InStock
75931-490
Anti-RBPMS Rabbit Polyclonal Antibody
Antibodies
RBPMS (RNA binding protein with multiple splicing), also known as HERMES, contains one RRM (RNA recognition motif) domain and belongs to the RRM family of RNA-binding proteins. RBPMS exists as multiple alternatively spliced isoforms and is thought to bind RNA, possibly playing a role in RNA-related events, such as transcription and translation. RNA-binding proteins that are specific to retinal ganglion cells (RGCs) have been previously identified as excellent markers for RGCs. Recent findings show that antibodies against RBPMS are robust reagents that exclusively identify RGCs in multiple mammalian species. RBPMS Antibody is ideal for researchers interested in neuroscience research.

Anti-RBPMS Antibody is stored with 0.03% sodium azide. It is suitable for Western Blots and Immunohistochemistry, and it is specific for the ~24 kDa RBPMS protein. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: RBPMS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty