Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RGS22 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DY2
75931-566EA 518 USD InStock
75931-566
Anti-RGS22 Rabbit Polyclonal Antibody
Antibodies
Regulator of G-protein signaling (RGS) proteins contain an 120 amino acid conserved domain, termed the RGS domain, that acts as a GTPase-activating protein that acts to reduce the signal transmitted by the receptor-activated G-alpha subunit. RGS22 is a recently identified member of this family that localizes to the testis and can interact with guanine nucleotide binding proteins alpha 11, 12, and 13 (GNA11, GNA12, and GNA13). While RGS22 has been postulated to play a role in spermiogenesis in the testis, it is also expressed in several cancer cell lines with an epithelial origin and associated with cancer metastasis. Its overexpression in a highly metastatic cancer causes a decrease in cell migration and a reduction of the invasive potential of the cells, suggesting that RGS22 may be a potential prognostic biomarker for metastasis.

Anti-RGS22 Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 147 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: RGS22
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty