Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HMMR Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DY3
75931-568EA 518 USD InStock
75931-568
Anti-HMMR Rabbit Polyclonal Antibody
Antibodies
The hyaluronan-mediated motility receptor, also known as RHAMM, was initially identified as a soluble protein that could be released by sub-confluent migrating cells, promoting cell motility and invasion via interactions with hyaluronan (HA) and the cell surface. While RHAMM is normally poorly expressed in most normal tissues and is not required for embryonic development or normal cell homeostasis functions, its expression is increased during wound repair in response to hypoxia and fibrogenic factors. However, its overexpression is transforming in multiple types of cancers and is required for maintaining RAS transformation. RHAMM associates with BRCA1 and BARD1, attenuating the mitotic-spindle-promoting activity of RHAMM, which may contribute to tumor progression by promoting genomic instability.

Anti-RHAMM Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 84 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: HMMR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty