Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RHOT1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DZ0
75931-598EA 518 USD InStock
75931-598
Anti-RHOT1 Rabbit Polyclonal Antibody
Antibodies
The Ras homolog family member T1 (RHOT) is an atypical Rho Ca2+-binding GTPase that localizes to the mitochondria (1). RHOT1, the related protein RHOT2, the adaptor protein Milton, and the PTEN induced putative kinase 1 (PINK1), form a complex that is involved in axonal transport of mitochondria (2,3). Both PINK1 and Parkin target RHOT1 for phosphorylation and degradation, causing the arrest of mitochondrial motility (4).

Anti-RHOT1 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 71 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: RHOT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty