Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RVFV_sL_gp1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DZ5
75931-616EA 518 USD InStock
75931-616
Anti-RVFV_sL_gp1 Rabbit Polyclonal Antibody
Antibodies
Rift Valley Fever (RFV) virus is an arthropod-borne virus endemic to Africa that infects humans and animals that is transmitted predominantly by mosquitoes (1). During human infections, symptoms can range from benign fever to severe encephalitis and fatal hepatitis with hemorrhagic fever. The Bunyaviridae family of viruses to which the RVF virus belongs are spherical enveloped viruses with a tripartite RNA genome of negative or ambisense polarity (2). The three segments are referred to as the L, M, and S segments. The L and M segments are negative polarity and code fore the L-dependent RNA polymerase and glycoprotein precursor respectively. The S segment is of ambisense polarity and encodes the nucleoprotein and non-structural proteins (3).

Anti-Rift Valley Fever Virus Polymerase Antibody has been tested for use in ELISA. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 238 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: RVFV_sL_gp1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty