Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RILPL1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DZ7
75931-622EA 518 USD InStock
75931-622
Anti-RILPL1 Rabbit Polyclonal Antibody
Antibodies
While the function of the Rab interacting lysosomal protein-like 1 (RILPL1) remains unknown, the homologous protein RILP is a small GTPase that controls transport to endocytic compartments, suggesting RILPL1 may have similar functions. However, overexpression of RILP caused enlarged lysosomes that are more centrally located in the cell, while overexpression of RILPL1 had no visible effect on the location or morphology of lysosomes.

Anti-RILPL1 Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 47 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: RILPL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty