Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RILPL2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-EA8
75931-638EA 518 USD InStock
75931-638
Anti-RILPL2 Rabbit Polyclonal Antibody
Antibodies
The Rab interacting lysosomal protein-like 2 (RLP2) protein is related to the is lysosomal protein RILP which interacts with RAB7, a small GTPase that controls transport to endocytic degradative compartments. Little is known of the exact function of RLP2, but it is thought that RILP is a downstream effector of RAB7 and both proteins act together in the regulation of late endocytic traffic, suggesting that RLP2 may be involved in endocytic traffic with RILP or play a similar role. RILP and possible RLP2 may also play a role in the biogenesis of multivesicular bodies.

Anti-RLP2 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 24 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: RILPL2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty