Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RNF168 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-EB1
75931-646EA 518 USD InStock
75931-646
Anti-RNF168 Rabbit Polyclonal Antibody
Antibodies
RNF168 was identified as a chromatin-associated RING finger protein that acts as a ubiquitin ligase both in vitro and in vivo. RNF168 targets histones H2A and H2AX, but not H2B, forming K63 polyubiquitin chains. Upon formation of DNA double strand breaks, RNF168 is recruited to the site of DNA damage where it co-localizes with gammaH2AX and 53BP1 in an RNF8-dependent manner. This localization of RNF168 increases the local concentration of ubiquinated proteins to the threshold required for retention of the proteins 53BP1 and BRCA1, facilitating the downstream signaling cascade. Thus, RNF168 defines a new pathway demonstrating a functional cooperation between E3 ligases in genome maintenance. At least three isoforms of RNF168 are known to exist.

Anti-RNF168 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 65 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: RNF168
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty