Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RNF8 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-EB3
75931-650EA 518 USD InStock
75931-650
Anti-RNF8 Rabbit Polyclonal Antibody
Antibodies
RNF8 was identified as a ubiquitin ligase (E3) containing a RING finger motif and a FHA domain. This protein has been shown to interact with several class II ubiquitin-conjugating enzymes including UBE2E1/UBCH6, UBE2E2, and UBE2E3. RNF8 assembles at DNA double-strand breaks (DSBs) via interactions though the FHA domain with the adaptor protein MDC1, resulting in an increase in DSB-associated H2A histone ubiquitinations mediated by the associated ubiquitin ligase RNF168 followed by the accumulation of 53BP1 and BRCA1 repair proteins. Together with RNF168, RNF8 plays an integral part of class switch recombination in B cells, allowing the production of several classes of antibodies, through the recruitment of 53BP1 and BRCA1 to the DSB sites.

Anti-RNF8 Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 56 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: RNF8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty