Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RSRC1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-EC4
75931-688EA 518 USD InStock
75931-688
Anti-RSRC1 Rabbit Polyclonal Antibody
Antibodies
The arginine/serine-rich coiled-coil 1 protein (RSRC1) was initially identified as a non-snRNP splicing factor that has a role in both the constitutive and alternative splicing of pre-mRNA molecules. In cellular nuclear extracts immunodepleted of RSRC1, the second step of pre-mRNA splicing is blocked; recombinant RSRC1 is able to restore splicing activity. RSRC1 has also been reported to be associated with schizophrenia as well as a unique marker of bone marrow-derived stem cells that also express the cdc34 marker.

Anti-RSRC1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 39 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: RSRC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty