Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SF3B4 Mouse Monoclonal Antibody [Clone: 3A1]

Supplier: Rockland Immunochemicals
200-301-X27
75931-722EA 528 USD InStock
75931-722
Anti-SF3B4 Mouse Monoclonal Antibody [Clone: 3A1]
Antibodies
The spliceosome associated protein of 49 kDa (SAP49), also known as SF3B4, is a widely expressed nuclear splicing factor that has been shown to have RNA binding activity as well as direct interaction with SAP145. The SAP145-SAP49 complex has recently been implicated in cell cycle progression as Vpr (viral protein 1 of HIV-1) has been shown to bind SAP145 thus interfering with the proper formation and functioning of the SAP145-SAP49 complex. SAP 49 Antibody is ideal for researchers interested in cell cycle research.

Anti-SAP 49 Antibody is suitable for Western Blots and Immunofluorescence, and it is specific for the ~49 kDa SAP 49 protein. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: SF3B4
Clonality: Monoclonal
Clone: 3A1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty