Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SCARB1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-EE0
75931-726EA 518 USD InStock
75931-726
Anti-SCARB1 Rabbit Polyclonal Antibody
Antibodies
Scavenger receptor class B member 1 (SCARB1), also known as SR-BI, is part of the scavenger receptor superfamily, which is composed of many members with diverse structures, expression patterns, and functions. SCARB1 is a multi-ligand cell-surface receptor that mediates the selective uptake of lipid from HDL cholesterol into cells and is expressed in steroidogenic tissues in adult animals. Other ligands of SCARB1 include native, acetylated, or oxidized LDL and anionic phospholipids. SCARB1-deficient mice have elevated HDL levels and increased susceptibility to atherosclerosis on fat feeding, indicating its importance in the regulation of cholesterol homeostasis. Along with CLDN1, LDL-R, and the tetraspanin superfamily member CD81, SCARB1 has been reported to be an entry factor for the Hepatitis C virus. At least two isoforms of SCARB1 are known to exist.

Anti-SCARB1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 61 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SCARB1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty