Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SCRN1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-EE6
75931-740EA 518 USD InStock
75931-740
Anti-SCRN1 Rabbit Polyclonal Antibody
Antibodies
SCRN1 was first identified as a cytosolic protein that is involved in the regulation of exocytosis from peritoneal mast cells. More recent studies have shown that SCRN1 expression is upregulated in gastric cancer cell lines and may possess epitopes that could function as tumor-associated antigens, potentially providing targets for cancer vaccines in the treatment of gastric cancers. Another report indicates that decreased expression of SCRN1 via RNAi expression resulted in significantly lower rates of cell growth in colorectal cancer cell lines, and increased SCRN1 expression in patients with colorectal cancer correlated with poor prognosis, suggesting that SCRN1 may also be involved in the regulation of cell growth and might be useful as a prognostic tool.

Anti-SCRN1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 46 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SCRN1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty