Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-B303_sMgp1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-EH1
75931-806EA 518 USD InStock
75931-806
Anti-B303_sMgp1 Rabbit Polyclonal Antibody
Antibodies
Severe fever with thrombocytopenia syndrome (SFTS) is an emerging infectious disease caused by SFTS virus, a newly discovered bunyavirus that can cause high rates of fatalities. This disease is thought to be transmitted through blood contact. The SFTS virus HB29 membrane glycoprotein polyprotein mRNA encodes two glycoproteins termed Phlebovirus glycoprotein G1 and G2 respectively. This antibody will detect the non-cleaved glycoprotein.

Anti-SFTS Virus HB29 Antibody has been tested for use in ELISA. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 117 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: B303_sMgp1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty