Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SHANK3 Mouse Monoclonal Antibody (ALP) [Clone: S367-51]

Supplier: Rockland Immunochemicals
200-305-W72
75931-814EA 539 USD InStock
75931-814
Anti-SHANK3 Mouse Monoclonal Antibody (ALP) [Clone: S367-51]
Antibodies
SHANK proteins are scaffolding adaptors that have been shown to integrate neurotransmitter receptors into the cortical cytoskeleton at postsynaptic densities. SHANK1-3 of the SHANK/ProSAP family are molecular scaffolds in the postsynaptic density (PSD). SHANK recruits betaPIX and PAK to dendritic spines to regulate postsynaptic structure and interacts with ionotropic receptor and metabotropic glutamate receptor complexes. Transcript splice variation in the Shank family influences the spectrum of Shank-interacting proteins in the PSDs of adult and developing brain to ensure normal development. Anti-SHANK1/SHANK3 is ideal for research in Neuroscience, including autism spectrum disorder, and Cell Adhesion.

Anti-SHANK1/SHANK3 Alkaline Phosphatase Conjugated Antibody is suitable for Western Blots, Immunohistochemistry and Immunocytochemistry. Expect a band approximately ~190kDa on specific lysates or tissues. It will cross react with SHANK1, but it will not cross react with SHANK2. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: SHANK3
Clonality: Monoclonal
Clone: S367-51
Conjugation: ALP
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty