Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SIGIRR Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-EK1
75931-876EA 518 USD InStock
75931-876
Anti-SIGIRR Rabbit Polyclonal Antibody
Antibodies
SIGIRR is a member of the Toll-like receptor-interleukin 1 receptor superfamily. Members of this family are defined by the presence of an intracellular Toll-IL-1R (TIR) domain. The Toll-like receptors (TLRs) are signaling molecules that recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses. SIGIRR was originally identified through database analysis and was shown to have only one Ig domain as opposed to the normal three Ig folds seen in the TIR family. Similar to ST2, another TIR family member, it has been shown to negatively regulate IL-1 receptor and Toll-like receptor signaling. However, SIGIRR inhibits TLR-IL-1R signaling by dimerizing with TLR4, TLR5, TLR9, and IL-1R. It also associates with the down-stream TLR signaling proteins IRAK and TRAF6 in an IL-1-dependent fashion.

Anti-SIGIRR Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 46 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SIGIRR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty