Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SIVA1 Chicken Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-901-EM3
75931-944EA 518 USD InStock
75931-944
Anti-SIVA1 Chicken Polyclonal Antibody
Antibodies
SIVA plays an important role in the apoptotic pathway induced by the CD27 antigen, a member of the tumor necrosis factor receptor (TFNR) superfamily. SIVA is a small cysteine-rich protein with a unique N-terminal amphipathic helical region, a death domain homology region (DDHR) and a C-terminal cysteine-rich B-box-like or zinc-finger-like structure. The DDHR domain binds to CD27 and induces apoptosis in T cells through a caspase-dependent mitochondrial pathway. SIVA inhibits BCL2L1 isoform Bcl-xL anti-apoptotic activity and inhibits activation of NFk-B and promotes T-cell receptor-mediated apoptosis. SIVA represents a potential therapeutic target for the treatment of neuronal cell death diseases.

Anti-SIVA Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 19 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SIVA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty