Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TTC37 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-W48
75931-956EA 518 USD InStock
75931-956
Anti-TTC37 Rabbit Polyclonal Antibody
Antibodies
SKI3 is a component of the SKI complex which is composed of SKI2, SKI3 and SKI8. The SKI complex interacts with SKI7, making the link between the SKI complex and the exosome in order to perform mRNA degradation. Anti-SKI3 Antibody is ideal for research in Gene Expression.

Anti-SKI3 Antibody is suitable for Chromatin Immunoprecipitation, Immunofluorescence and Western Blots. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 150 kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: TTC37
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty