Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SLC35D3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-EP5
75931-990EA 518 USD InStock
75931-990
Anti-SLC35D3 Rabbit Polyclonal Antibody
Antibodies
The solute carrier family SLC35 consists of at least 17 proteins that act as nucleotide sugar transporters localized to the Golgi apparatus and endoplasmic reticulum. The novel protein SLC35D3 is highly homologous to SLC35D1 and SLC35D2, both of which transport UDP-glucuronic acid and UDP-N-acetylgalactosamine, suggesting that SLC35D3 is also involved in the transport of nucleotide sugars. It has been suggested that SLC35D3 regulates platelet dense granules, lysosome-related organelles which contain high concentrations of several biologically important low molecular weight molecules necessary for normal blood homeostasis.

Anti-SLC35D3 Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 44 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SLC35D3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty