Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SLITRK2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-EQ1
75932-002EA 518 USD InStock
75932-002
Anti-SLITRK2 Rabbit Polyclonal Antibody
Antibodies
SLIT and NTRK-like family 2 (Slitrk2) is a member a protein family consisting of six homologous transmembrane proteins (Slitrk1-6) that share two conserved leucine-rich repeat domains in the extracellular domain and have significant homology to Slit, a secreted axonal growth-controlling protein. These proteins are also homologous to trk neurotrophin receptors in their intracellular domains. Expression of Slitrk proteins is highly restricted to neural and brain tumor tissues, but varies within the family. For example, Slitrk2 is expressed primarily in neurons in the ventricular layer of the brain. Like every other Slitrk protein except Slitrk1, overexpression of Slitrk2 inhibited neurite outgrowth in cultured neurons. Deletion analysis indicated that the functional difference between Slitrk2 and Slitrk1 is within their intracellular domains. At least two isoforms of this protein are known to exist.

Anti-Slitrk2 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 95 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SLITRK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty