Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SNAP25 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X28
75932-066EA 530 USD InStock
75932-066
Anti-SNAP25 Rabbit Polyclonal Antibody
Antibodies
SNAP25 (Synaptosomal associated protein of 25 kDa) is a presynaptic plasma membrane protein that is widely distributed throughout the brain and involved in the regulation of neurotransmitter release. Decreased levels of SNAP25 have been found in the brains of patients with Down Syndrome and Alzheimer’s Disease. In addition, a significant reduction in the hippocampal expression of SNAP25 has also been found in patients with Schizophrenia. Increasing evidence suggests that SNAP-25 also modulates various ion channels, including voltage gated calcium channels (VGCCs). Activation of PKC results in the phosphorylation of SNAP-25 on ser187. Phosphorylation of SNAP25 on ser187 is believed to cause inhibition of VGCC. Since ser187 phosphorylation is transiently induced by neuronal activity, SNAP25 creates a negative feedback mechanism for controlling neuronal excitability. Olig2 pS 10/13/14 Antibody is ideal for researchers interested in neuroscience research. SNAP25 pS187 Antibody is ideal for researchers interested in neuroscience research.

Anti-Phospho-Ser187 SNAP25 Antibody is stored with 100 μg per ml BSA. It is suitable for Western Blots and is specific for the ~25 kDa SNAP25 protein phosphorylated at Ser187 in Western blots. Immunolabeling is completely blocked by blocked lambda-Phosphatase. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: SNAP25
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty