Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BAG4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-ES1
75932-096EA 518 USD InStock
75932-096
Anti-BAG4 Rabbit Polyclonal Antibody
Antibodies
Apoptosis is induced by certain cytokines including TNF and Fas ligand of the TNF family through their death domain containing receptors, TNF-R1 and Fas. Several novel death receptors including DR3, DR4, DR5, and DR6 were recently identified. Cell death signal is transduced by death domain containing adapter molecules through the interaction with death domain of these death receptors. A novel TNF-R1 interacting protein was recently identified and designated SODD for silencer of death domains. SODD associates with the death domain of TNF-R1 and prevents constitutive activation of TNF-R1 signaling. TNF treatment releases SODD and permits adapter molecules such as TRADD recruiting to the active TNF-R1 complex, which activates TNF signaling pathways. SODD also interacts with DR3. SODD is ubiquitously expressed in human tissues and cell lines.

Anti-SODD Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 50 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: BAG4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty