Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SOX17 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-ES3
75932-118EA 518 USD InStock
75932-118
Anti-SOX17 Rabbit Polyclonal Antibody
Antibodies
SOX17 is a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate (1,2). SOX17 is part of the SoxF subgroup which plays an important role in the differentiation of different cell types, including visceral and definitive endoderm and hematopoietic cell types (3,4). SOX17 is also essential for acquisition and maintenance of arterial identity (5).

Anti-SOX17 Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 44 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SOX17
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty