Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SOX18 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-ES4
75932-120EA 518 USD InStock
75932-120
Anti-SOX18 Rabbit Polyclonal Antibody
Antibodies
SOX18 is a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate (1,2). SOX18 is part of the SoxF subgroup which plays an important role in the differentiation of different cell types (3). SOX is known to be involved with vascularization and endothelial development, suggesting that it may be useful as a potential target for inhibiting tumor angiogenesis (4). Mutations in this gene have been associated with recessive and dominant forms of hypotrichosis-lymphedema-telangiectasia (5).

Anti-SOX18 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 41 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SOX18
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty