Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SFTPA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-R06
75932-128EA 487 USD InStock
75932-128
Anti-SFTPA1 Rabbit Polyclonal Antibody
Antibodies
Surfactant protein A is an innate immune system collectin. The gene encoding SP-A map to mouse chromosome 14. Surfactant protein-A, which plays a role in innate host defense in the lung, is also expressed in the Eustachian tube. We report that the frequency of specific surfactant protein-A haplotypes and genotypes differs between children with recurrent otitis media compared with a control population. This antibody is suitable for researchers interested in cancer research.

Anti-SP-A is useful for Immunohistochemistry -P and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~26.2 kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: SFTPA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty