Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SPINK2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-ET4
75932-148EA 518 USD InStock
75932-148
Anti-SPINK2 Rabbit Polyclonal Antibody
Antibodies
Human serine proteinase inhibitor Kazal-type 2 (SPINK2) is required for maintaining normal spermatogenesis and potentially regulates serine protease-mediated apoptosis in male germ cells. It contains a typical kazal domain composed by six cysteine residues forming three disulfide bridges. SPINK2 functions as a trypsin/acrosin inhibitor and is synthesized mainly in the testis and seminal vesicle where its activity is engaged in fertility. SPINK2 plays a role in the pathogenesis of hereditary and chronic pancreatitis.

Anti-SPINK2 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 9 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SPINK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty