Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SQSTM1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-EU6
75932-160EA 518 USD InStock
75932-160
Anti-SQSTM1 Rabbit Polyclonal Antibody
Antibodies
SQSTM1/p62 is an adapter protein which binds ubiquitin and regulates signaling cascades through ubiquitination. It may regulate the activation of NF-kB by TNF-a, nerve growth factor (NGF) and interleukin-1. SQSTM1/p62, a co-interacting protein of the atypical PKC isoforms, has a UBA domain at its C-terminal end, which binds non-covalently to polyubiquitin chain. SQSTM1's UBA domain is necessary for recruitment of polyubiquitin and aggresome formation. SQSTM1 may play a role in titin/TTN downstream signaling in muscle cells and may be involved in cell differentiation, apoptosis, immune response and regulation of K+ channels. Mutations in the ubiquitin-associated (UBA) domain of SQSTM1 commonly cause Paget's disease of bone since the UBA is necessary for aggregate sequestration and cell survival.

Anti-SQSTM1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry, and Immunofluorescence. Expect a band at approximately 48 kDa in Western Blots using positive control Human spleen tissue, rat spleen tissue for IHC and IF, or other specific cell lysates and tissues. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: SQSTM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty