Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Il1rl1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-H20
75932-174EA 518 USD InStock
75932-174
Anti-Il1rl1 Rabbit Polyclonal Antibody
Antibodies
ST2 antibody detects mouse ST2 and is common to all three isoforms. ST2 is a member of a superfamily containing the interleukin-1 receptor and the Toll-like receptors (TLRs). The TLRs are signaling molecules that recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses. ST2 was originally identified as a protein whose production was stimulated by various proliferation-inducing agents such as PDGF and FGF. More recently, it has been shown to negatively regulate IL-1 receptor and Toll-like receptor (TLR) 4 signaling and to maintain endotoxin tolerance. It has been suggested that the inhibition of TLR4 signaling occurs through the association and sequestering of TLR adaptor molecules such as MyD88 and TIRAP. Anti-ST2 antibodies are ideal for investigators involved in cytokine and growth factors research.

Anti-ST2 Antibody is suitable for use in E,WB, and IHC. Expect a band approximately ~64.8kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: Il1rl1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty