Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STAT1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-EV3
75932-178EA 518 USD InStock
75932-178
Anti-STAT1 Rabbit Polyclonal Antibody
Antibodies
STATs (signal transducers and activators of transcription) are a family of cytoplasmic latent transcription factors that are activated to regulate gene expression in response to a large number of extracellular signaling polypeptides including cytokines, interferons, and growth factors. After phosphorylation by JAK tyrosine kinases, STATs enter the nucleus to regulate transcription of many different genes. Among the seven STATs (Stat1, Stat2, Stat3, Stat4, Stat5a, Stat5b, and Stat6), Stat1, Stat3, Stat5a, and Stat5b have a wide activation profile. STAT1 is activated by many different ligands including IFN family (IFN-a, IFN-b, IFN-gamma and IL-10), gp130 family (IL-6, IL-11, LIF, CNTF, and G-CSF), and receptor tyrosine kinases (EGF, PDGF, and CSF-1). STAT1 has two forms, the 91 kDa STAT1a and the 84 kDa STAT1b which are encoded by the same gene with splicing variant.

Anti-STAT1 alpha Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry and Immunoprecipitation. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 87 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: STAT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty