Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STAT1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GH1
75932-180EA 518 USD InStock
75932-180
Anti-STAT1 Rabbit Polyclonal Antibody
Antibodies
Signal transducer and activator of transcription 1 (Stat1) belongs to a family of cytoplasmic transcription factors that can be activated by a cell surface receptor. Stat1 has two isoforms. Stat1 has been found to be inappropriately activated in many tumors. In addition to tyrosine phosphorylation, Stat1 is phosphorylated through a p38 mitogen-activated protein kinase (MAPK)-dependent pathway at Ser727 in response to IFN-g and other cellular stresses. Stat1 is localized in the cytoplasm and upon phosphorylation at Y701 is translocated to the nucleus.

Anti-STAT1 [Asym-dimethyl Arg31] antibody is useful for Western Blot and Dot Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~87 kDa corresponding to STAT1 protein by Western Blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: STAT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty