Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STAT3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GH6
75932-192EA 518 USD InStock
75932-192
Anti-STAT3 Rabbit Polyclonal Antibody
Antibodies
Signal transducer and activator of transcription 3 (Stat3) belongs to a family of cytoplasmic transcription factors that can be activated by phosphorylation by its cell surface receptor. Stat3 plays a key role in many cellular processes such as cell growth and apoptosis. It also mediates cellular responses to interleukins, KITLG/SCF,EGF, IFN-alpha and other growth factors and may mediate cellular responses to activated FGFR1, FGFR2, FGFR3 and FGFR4. Stat3 forms a homodimer or a heterodimer with a related family member (e.g. STAT1). Activation occurs through phosphorylation of tyrosine 705 and serine 727. Phosphorylation of Stat3 at Tyr705 induces Stat3 dimerization and nuclear translocation. Serine phosphorylation is important for stable DNA-binding of Stat3 homodimers and maximal transcriptional activity. Stat3 can have a dual role in cancer, it has been found that Stat3 protein can promote oncogenesis and have a tumor suppressor role depending upon the mutational background of the tumor.

Anti-STAT3 antibody is useful for Western Blot and Dot Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~88 kDa corresponding to STAT3 protein by Western Blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: STAT3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty