Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ycjM Goat Polyclonal Antibody (BAC)

Supplier: Rockland Immunochemicals
200-106-231-0100
75932-370EA 498 USD InStock
75932-370
Anti-ycjM Goat Polyclonal Antibody (BAC)
Antibodies
Anti-Sucrose Phosphorylase recognizes sucrose phosphorylase, a member of the hexosyltransferases family of enzymes. Sucrose phosphylase catalyzes the conversion of sucrose to fructose and glucose-1-phosphate both of which can be used in glycolysis, gluconeogenesis, and pentose phosphate pathway.

Anti-Sucrose Phosphorylase is suitable for investigators interested in Metabolism, Cancer, and Signal Transduction.

Anti-Sucrose Phosphorylase antibody has been assayed against 1.0 µg of Sucrose Phosphorylase in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature.  A working dilution of 1:25,000 to 1:115,000 of the reconstitution concentration is suggested for this product.

Type: Primary
Antigen: ycjM
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Goat
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty