Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NP Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-EX8
75932-410EA 518 USD InStock
75932-410
Anti-NP Rabbit Polyclonal Antibody
Antibodies
Influenza A virus is a major public health threat, killing more than 30, 000 people per year in the USA. In early 2009, a novel swine-origin influenza A (H1N1) virus (S-OIV) was identified in specimens obtained from patients in Mexico and the United States. The influenza A virus polymerase transcribes and replicates eight virion RNA (vRNA) segments, among which the nucleocapsid protein (NP), thought to control whether mRNA or cRNA is produced. The nucleoprotein (NP), which has multiple functions during the virus life cycle, possesses regions that are highly conserved among influenza A, B, and C viruses. It was recently found several NP mutations that affected the efficient incorporation of multiple viral-RNA (vRNA) segments into progeny virions even though a single vRNA segment was incorporated efficiently. This indicates that the respective conserved amino acids in NP may be critical for the assembly and/or incorporation of sets of eight vRNA segments.

Anti-Swine H1N1 Nucleocapsid Protein Antibody has been tested for use in ELISA and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 56 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty