Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Rabbit Polyclonal Antibody (DL800)

Supplier: Rockland Immunochemicals
614-445-002
75932-412EA 130 USD InStock
75932-412
Anti-IgG Rabbit Polyclonal Antibody (DL800)
Antibodies
SWINE IgG (H&L) (RABBIT) Antibody generated in rabbit detects specifically swine IgG whole molecule. Anti-Swine IgG antibody is ideal for investigators involved in serum rotein component research.

Anti-SWINE IgG (H&L) (RABBIT) Antibody is suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunohistochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency. The emission spectra for this DyLight™ conjugate match the principle output wavelengths of most common fluorescence instrumentation. Specific condtions should be optimized by user.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DL800
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pig

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty