Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TLN1, TLN2 Mouse Monoclonal Antibody [Clone: Tal-33]

Supplier: Rockland Immunochemicals
200-301-P47
75932-568EA 528 USD InStock
75932-568
Anti-TLN1, TLN2 Mouse Monoclonal Antibody [Clone: Tal-33]
Antibodies
Talin is a high-molecular-weight cytoskeletal protein concentrated at regions of cell-substratum contact and, in lymphocytes, at cell-cell contacts. Talin serves a function of linking vinculin to the integrins, and, thus, the cytoskeleton to extracellular matrix (ECM) receptors. It has a mass of 270 kD and shares 23% N-terminal identity with ezrin, which has similar functions. TLN gene is mapped to 9p. This antibody is suitable for researchers interested in cell signaling, membrane biology, and growth factor research.

Anti-Talin is useful for Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~270-272 kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: TLN1, TLN2
Clonality: Monoclonal
Clone: Tal-33
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty