Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TASK1 Mouse Monoclonal Antibody (ALP) [Clone: S374-48]

Supplier: Rockland Immunochemicals
200-305-W85
75932-582EA 539 USD InStock
75932-582
Anti-TASK1 Mouse Monoclonal Antibody (ALP) [Clone: S374-48]
Antibodies
K+ channels are divided into three subclasses reflecting the number of transmembrane segments (TMS), which are designated 6TMS, 4TMS and 2TMS. Members of the 4TMS class contain two distinct pore regions and include TWIK, TREK, TRAAK and TASK. TASK channels are highly sensitive to external pH in the physiological range. TASK-1 is expressed in brain and in rat heart, with high levels of expression in the right atrium. TASK-2, mainly expressed in kidney, is localized in cortical distal tubules and collecting ducts, suggesting a role in renal K+ transport. TASK-3 from rat cerebellum shares 54% identity with TASK-1, but less than 30% identity with TASK-2 and other tandem pore K+ channels. Anti-TASK1 Potassium Channel Antibody is ideal for research in Neuroscience, particularly studies concerning the dopaminergic neuron and melatonin signaling.

Anti-TASK1 Potassium Channel Alkaline Phosphatase Conjugated Antibody is suitable for Western Blots, Immunohistochemistry and Immunocytochemistry. Expect a band approximately ~50kDa on specific lysates or tissues. Does not cross react with TASK3. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: TASK1
Clonality: Monoclonal
Clone: S374-48
Conjugation: ALP
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty