Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TASP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-V93
75932-616EA 487 USD InStock
75932-616
Anti-TASP1 Rabbit Polyclonal Antibody
Antibodies
TASP1 is an endopeptidase which cleaves specific substrates after an aspartate residue. It is required for the posttranslational processing of MLL, a histone methyl transferase which is essential for the maintenance of HOX gene expression and for the transcription factor TFIIA. Therefore, TASP1 may have an important function in the regulation of gene transcription and embryonic development. Anti-TASP1 Antibody is ideal for research in Cell Differentiation and Development, Gene Expression and Epigenetics.

Anti-TASP1 Antibody is suitable for ELISA and Western Blots. Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 44.5 kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: TASP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty