Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAPT Chicken Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-901-X29
75932-618EA 518 USD InStock
75932-618
Anti-MAPT Chicken Polyclonal Antibody
Antibodies
Tau is a key microtubule-associated protein that plays an important role in the formation of microtubules in axons. Six tau isoforms have been identified as products of a single gene produced by alternative mRNA splicing. Tau mutations have been implicated in many neurodegenerative disorders such as Alzheimer’s disease (AD), Pick’s disease and progressive supranuclear palsy. Tau Antibody is ideal for researchers interested in neuroscience research.

Anti-Tau Antibody is suitable for Western Blots and Immunofluorescence, and it is specific for the ~48, 65 & 75 kDa tau isoforms. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: MAPT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty