Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TDRG1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FB5
75932-800EA 518 USD InStock
75932-800
Anti-TDRG1 Rabbit Polyclonal Antibody
Antibodies
The Testis development related protein 1 (TDRG1) is thought to be a testis-specific, developmentally regulated gene. TDRG1 is expressed in mainly in spermatogenic cells in seminiferous tubules of the adult testis, but is not detected in fetal testis. While little is known of the function of TDRG1, it has been postulated to play an important role in human spermatogenesis.

Anti-TDRG1 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 10 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TDRG1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty