Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TEKT5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FB6
75932-802EA 518 USD InStock
75932-802
Anti-TEKT5 Rabbit Polyclonal Antibody
Antibodies
Tektins are insoluble alpha-helical proteins related to intermediate filament proteins and are essential for the assembly of cilia and flagella. Tektin-5 (TEKT5) is a recently identified member of the Tektin family. It is predominantly expressed in testis, and its expression is upregulated during testis development. It is present in sperm flagella but not in heads, suggesting that it may act as a middle component required for flagellar stability and sperm mobility.

Anti-TEKT5 Antibody has been tested for use in ELISA, Western Blotting, and Immunohistochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 56 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TEKT5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty