Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNN Mouse Monoclonal Antibody [Clone: T20]

Supplier: Rockland Immunochemicals
200-301-P48
75932-822EA 528 USD InStock
75932-822
Anti-TNN Mouse Monoclonal Antibody [Clone: T20]
Antibodies
The tenascins are a family of extracellular matrix proteins (ECMs). Tenascin is an extracellular matrix protein. It expressed in an unusually restricted pattern during embryogenesis and has been implicated in a variety of morphogenetic phenomena. Human hexabrachion gene(tenascin) is mapped to chromosome 9, bands q32-q34. The coding region of the hexabrachion gene spans approximately 80 kilobases of DNA and consists of 27 exons separated by 26 introns. This antibody is suitable for researchers interested in extracellular matrix antibodies.

Anti-Tenascin is useful for Immunohistochemistry -P, -F, Immunocytochemistry, and Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~144 kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: TNN
Clonality: Monoclonal
Clone: T20
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty