Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TGM5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FD5
75932-852EA 518 USD InStock
75932-852
Anti-TGM5 Rabbit Polyclonal Antibody
Antibodies
Transglutaminases (TGM) are a family of structurally and functionally related Ca2+-dependent enzymes (TGases) that stabilize protein assemblies through the formation of gamma-glutamyl-epsilon lysine crosslinks. TGases influence numerous biological processes, including blood coagulation, cell differentiation, fertilization and apoptosis. TGM5 belongs to the transglutaminase superfamily and catalyzes the cross-linking of proteins between glutamine and lysine residues, often resulting in stabilization of protein assemblies. TGM5 is also expressed in the human hair follicle. Defects in TGM5 are associated with acral peeling skin syndrome.

Anti-TGM5 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 81 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TGM5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty