Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-THAP11 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FD7
75932-856EA 518 USD InStock
75932-856
Anti-THAP11 Rabbit Polyclonal Antibody
Antibodies
The THAP domain-contining protein 11 (THAP11), also known as Ronin, is an essential factor involved in embryonic stem (ES) cell pluripotency and cell growth (1). THAP 11 contains a THAP domain, a conserved DNA-binding domain common to many proteins associated with chromatin modification and gene expression silencing, and has striking similarity to the site-specific DNA-binding domain (DBD) of Drosophila P element transposases (2). THAP11 can also negatively regulate CD44 v6 expression through its interaction with the poly(rC) binding protein PCBP1 (3).

Anti-THAP11 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 34 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: THAP11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty