Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-THBS2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-R01
75932-858EA 487 USD InStock
75932-858
Anti-THBS2 Rabbit Polyclonal Antibody
Antibodies
Thrombospondin-2 (THBS2) is a protein that in humans is encoded by the THBS2 gene. The protein encoded by this gene belongs to the thrombospondin family. The THBS2 is mapped to 6q27 and it is located on chromosome 17. The gene was transcribed in fibroblasts, smooth muscle cells, and an osteosarcoma cell line. It functions as a protein inhibitor of tumor growth and angiogenesis and modulates the cell surface properties of mesenchymal cells and be involved in cell adhesion and migration. This antibody is suitable for researchers interested in cancer research.

Anti-THBS2 is useful for Immunohistochemistry -P and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~130 kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: THBS2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty