Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TMPO Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FE1
75932-898EA 518 USD InStock
75932-898
Anti-TMPO Rabbit Polyclonal Antibody
Antibodies
Thymopoietin (TMPO), also known as LAP2, is a widely expressed lem-domain-containing nuclear protein that is thought to play an important role in the regulation of nuclear architecture by binding lamin B1 and chromosomes in a manner regulated by phosphorylation during mitosis. Thymopoietin associates with nucleoplasmic lamins A/C, which are encoded by the dilated cardiomyopathy (DCM) gene LMNA; mutations in Thymopoietin disrupt this binding, suggesting that Thymopoietin may be a candidate disease gene for DCM.

Anti-Thymopoietin Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 75 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TMPO
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty